Assay Details
| Gene Symbol: F26F4.9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065630.6 | NP_498031.3 | 2 - 3 | 253 | 74 | |
| Gene Symbol: F26F4.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065629.5 | NP_498030.1 | 2 - 3 | 233 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_065629.5 transcripts.
Set Membership: |
|

F26F4.9
Genomic Map