Assay Details
| Gene Symbol: ula-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361710.4 | NP_001348740.1 | 2 - 3 | 449 | 90 | |
| NM_065636.5 | NP_498037.2 | 2 - 3 | 447 | 90 | ||
| Gene Symbol: mmab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384085.1 | 6 - 7 | 1478 | 90 | ||
More Information
Set Membership: |
|

ula-1
Genomic Map