Assay Details
| Gene Symbol: mrps-18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065644.6 | NP_498045.2 | 2 - 3 | 578 | 68 | |
| Gene Symbol: fsn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065645.7 | NP_498046.1 | 2 - 3 | 580 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_065645.7 transcripts.
Set Membership: |
|

mrps-18C
Genomic Map