Assay Details
| Gene Symbol: wdr-60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381848.1 | NP_001367569.1 | 1 - 2 | 96 | 144 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdr-60| Gene Symbol: wdr-60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381848.1 | NP_001367569.1 | 1 - 2 | 96 | 144 | |
Set Membership: |
|