Assay Details
| Gene Symbol: rbm-42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065689.4 | NP_498090.1 | 2 - 3 | 438 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rbm-42| Gene Symbol: rbm-42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065689.4 | NP_498090.1 | 2 - 3 | 438 | 76 | |
Set Membership: |
|