Assay Details
| Gene Symbol: cid-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065698.6 | NP_498099.1 | 8 - 9 | 3578 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cid-1| Gene Symbol: cid-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065698.6 | NP_498099.1 | 8 - 9 | 3578 | 83 | |
Set Membership: |
|