Assay Details
| Gene Symbol: hira-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356742.3 | NP_001343798.1 | 2 - 3 | 1296 | 127 | |
| NM_065700.6 | NP_498101.2 | 10 - 11 | 2613 | 127 | ||
| NR_151567.1 | 9 - 10 | 2913 | 127 | |||
More Information
Set Membership: |
|

hira-1
Genomic Map