Assay Details
| Gene Symbol: nex-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065708.9 | NP_498109.1 | 2 - 3 | 580 | 71 | |
| GenBank mRNA | U40483.1 | - | 1 - 2 | 515 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nex-1| Gene Symbol: nex-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065708.9 | NP_498109.1 | 2 - 3 | 580 | 71 | |
| GenBank mRNA | U40483.1 | - | 1 - 2 | 515 | 71 | |
Set Membership: |
|