Assay Details
| Gene Symbol: prmt-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065711.8 | NP_498112.1 | 1 - 2 | 307 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prmt-5| Gene Symbol: prmt-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065711.8 | NP_498112.1 | 1 - 2 | 307 | 67 | |
Set Membership: |
|