Assay Details
| Gene Symbol: ztf-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065723.6 | NP_498124.2 | 8 - 9 | 1285 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ztf-8| Gene Symbol: ztf-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065723.6 | NP_498124.2 | 8 - 9 | 1285 | 72 | |
Set Membership: |
|