Assay Details
| Gene Symbol: glrx-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047426.3 | NP_001040891.1 | 2 - 3 | 260 | 127 | |
| NM_001047427.4 | NP_001040892.1 | 1 - 2 | 176 | 127 | ||
More Information
Set Membership: |
|

glrx-21
Genomic Map