Assay Details
| Gene Symbol: thoc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267997.2 | NP_001254926.1 | 1 - 2 | 132 | 68 | |
| NM_001267998.4 | NP_001254927.1 | 1 - 2 | 166 | 68 | ||
More Information
Set Membership: |
|

thoc-5
Genomic Map