Assay Details
| Gene Symbol: thoc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267997.2 | NP_001254926.1 | 6 - 7 | 1590 | 85 | |
| NM_001267998.4 | NP_001254927.1 | 6 - 7 | 1630 | 85 | ||
More Information
Set Membership: |
|

thoc-5
Genomic Map