Assay Details
| Gene Symbol: paqr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065747.7 | NP_498148.2 | 5 - 6 | 521 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

paqr-2| Gene Symbol: paqr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065747.7 | NP_498148.2 | 5 - 6 | 521 | 77 | |
Set Membership: |
|