Assay Details
| Gene Symbol: paa-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065761.5 | NP_498162.2 | 3 - 4 | 1601 | 91 | |
| GenBank mRNA | AB110419.1 | - | 3 - 4 | 1590 | 91 | |
More Information
Set Membership: |
|

paa-1
Genomic Map