Assay Details
| Gene Symbol: rcan-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065822.7 | NP_498223.2 | 1 - 2 | 184 | 131 | |
| GenBank mRNA | AF176115.1 | - | 1 - 2 | 161 | 131 | |
More Information
Set Membership: |
|

rcan-1
Genomic Map