Assay Details
| Gene Symbol: rgs-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129200.4 | NP_001122672.1 | 1 - 2 | 189 | 75 | |
| NM_065828.8 | NP_498229.3 | 1 - 2 | 314 | 75 | ||
More Information
Set Membership: |
|

rgs-5
Genomic Map