Assay Details
| Gene Symbol: rbm-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361715.4 | NP_001348727.1 | 4 - 5 | 814 | 84 | |
| NM_065833.5 | NP_498234.1 | 4 - 5 | 808 | 84 | ||
More Information
Set Membership: |
|

rbm-26
Genomic Map