Assay Details
| Gene Symbol: ida-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065844.6 | NP_498245.2 | 8 - 9 | 1699 | 76 | |
| GenBank mRNA | AF126740.1 | - | 8 - 9 | 1656 | 76 | |
| AJ245560.1 | - | 8 - 9 | 1656 | 76 | ||
More Information
Set Membership: |
|

ida-1
Genomic Map