Assay Details
| Gene Symbol: ddx-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065859.6 | NP_498260.2 | 4 - 5 | 2000 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddx-23| Gene Symbol: ddx-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065859.6 | NP_498260.2 | 4 - 5 | 2000 | 126 | |
Set Membership: |
|