Assay Details
| Gene Symbol: mlp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276776.3 | NP_001263705.1 | 1 - 2 | 112 | 62 | |
| NM_001372606.3 | NP_001360006.1 | 2 | 154 | 62 | ||
| NM_065899.7 | NP_498300.2 | 2 - 3 | 191 | 62 | ||
More Information
Set Membership: |
|

mlp-1
Genomic Map