Assay Details
| Gene Symbol: mvk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027695.4 | NP_001022866.1 | 2 - 3 | 320 | 109 | |
| NM_001027696.7 | NP_001022867.1 | 2 - 3 | 485 | 109 | ||
| NM_001379745.1 | NP_001367850.1 | 2 - 3 | 183 | 109 | ||
| NM_001379746.2 | NP_001367849.1 | 2 - 3 | 322 | 109 | ||
More Information
Set Membership: |
|

mvk-1
Genomic Map