Assay Details
| Gene Symbol: rpn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065945.6 | NP_498346.2 | 1 - 2 | 88 | 52 | |
| NM_065946.7 | NP_498347.3 | 2 - 3 | 85 | 52 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpn-2| Gene Symbol: rpn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065945.6 | NP_498346.2 | 1 - 2 | 88 | 52 | |
| NM_065946.7 | NP_498347.3 | 2 - 3 | 85 | 52 | ||
Set Membership: |
|