Assay Details
| Gene Symbol: rpn-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065945.6 | NP_498346.2 | 5 - 6 | 2121 | 109 | |
| NM_065946.7 | NP_498347.3 | 6 - 7 | 2118 | 109 | ||
More Information
Set Membership: |
|

rpn-2
Genomic Map