Assay Details
| Gene Symbol: thoc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065991.7 | NP_498392.2 | 12 - 13 | 3722 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

thoc-2| Gene Symbol: thoc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065991.7 | NP_498392.2 | 12 - 13 | 3722 | 151 | |
Set Membership: |
|