Assay Details
| Gene Symbol: zip-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066025.4 | NP_498426.1 | 3 - 4 | 412 | 78 | |
| NM_181936.6 | NP_871665.1 | 2 - 3 | 635 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zip-8| Gene Symbol: zip-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066025.4 | NP_498426.1 | 3 - 4 | 412 | 78 | |
| NM_181936.6 | NP_871665.1 | 2 - 3 | 635 | 78 | ||
Set Membership: |
|