Assay Details
| Gene Symbol: ddo-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066052.5 | NP_498453.1 | 3 - 4 | 793 | 88 | |
| GenBank mRNA | AB275893.1 | - | 3 - 4 | 760 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddo-3| Gene Symbol: ddo-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066052.5 | NP_498453.1 | 3 - 4 | 793 | 88 | |
| GenBank mRNA | AB275893.1 | - | 3 - 4 | 760 | 88 | |
Set Membership: |
|