Assay Details
| Gene Symbol: rbf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027395.5 | NP_001022566.1 | 2 - 3 | 305 | 101 | |
| NM_001027396.5 | NP_001022567.1 | 2 - 3 | 107 | 101 | ||
| NM_001379772.1 | NP_001367825.1 | 4 - 5 | 818 | 101 | ||
| NM_066066.7 | NP_498467.3 | 4 - 5 | 781 | 101 | ||
| GenBank mRNA | AF399852.1 | - | 1 - 2 | 85 | 101 | |
More Information
Set Membership: |
|

rbf-1
Genomic Map