Assay Details
| Gene Symbol: rbf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027395.5 | NP_001022566.1 | 6 - 7 | 1031 | 100 | |
| NM_001027396.5 | NP_001022567.1 | 6 - 7 | 833 | 100 | ||
| NM_001379772.1 | NP_001367825.1 | 8 - 9 | 1544 | 100 | ||
| NM_066066.7 | NP_498467.3 | 8 - 9 | 1507 | 100 | ||
| GenBank mRNA | AF399852.1 | - | 5 - 6 | 811 | 100 | |
More Information
Set Membership: |
|

rbf-1
Genomic Map