Assay Details
| Gene Symbol: rbf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027395.5 | NP_001022566.1 | 8 - 9 | 1176 | 69 | |
| NM_001027396.5 | NP_001022567.1 | 8 - 9 | 978 | 69 | ||
| NM_001379772.1 | NP_001367825.1 | 10 - 11 | 1689 | 69 | ||
| NM_066066.7 | NP_498467.3 | 10 - 11 | 1652 | 69 | ||
| GenBank mRNA | AF399852.1 | - | 7 - 8 | 956 | 69 | |
More Information
Set Membership: |
|

rbf-1
Genomic Map