Assay Details
| Gene Symbol: ten-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027552.5 | NP_001022723.2 | 2 - 3 | 216 | 78 | |
| NM_001268011.3 | NP_001254940.1 | 5 - 6 | 610 | 78 | ||
| NM_001268012.3 | NP_001254941.1 | 3 - 4 | 388 | 78 | ||
| NM_001379777.2 | NP_001367791.1 | 2 - 3 | 222 | 78 | ||
| GenBank mRNA | AB206835.1 | - | 2 - 3 | 216 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001379777.2 transcripts.
Set Membership: |
|

ten-1
Genomic Map