Assay Details
| Gene Symbol: zak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372611.2 | NP_001360014.1 | 3 - 4 | 771 | 75 | |
| NM_001379778.1 | NP_001367790.1 | 3 - 4 | 789 | 75 | ||
| NM_001379779.2 | NP_001367792.1 | 3 - 4 | 794 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001372611.2 transcripts.
Set Membership: |
|

zak-1
Genomic Map