Assay Details
| Gene Symbol: cra-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066093.6 | NP_498494.2 | 15 - 16 | 1955 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cra-1| Gene Symbol: cra-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066093.6 | NP_498494.2 | 15 - 16 | 1955 | 88 | |
Set Membership: |
|