Assay Details
| Gene Symbol: prk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276777.3 | NP_001263706.1 | 8 - 9 | 1375 | 70 | |
| NM_001377564.1 | NP_001364690.1 | 7 - 8 | 1118 | 70 | ||
More Information
Set Membership: |
|

prk-1
Genomic Map