Assay Details
| Gene Symbol: prk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276777.3 | NP_001263706.1 | 9 - 10 | 1542 | 63 | |
| NM_001377564.1 | NP_001364690.1 | 8 - 9 | 1285 | 63 | ||
More Information
Set Membership: |
|

prk-1
Genomic Map