Assay Details
| Gene Symbol: prk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276777.3 | NP_001263706.1 | 10 - 11 | 1673 | 106 | |
| NM_001377564.1 | NP_001364690.1 | 9 - 10 | 1416 | 106 | ||
More Information
Set Membership: |
|

prk-1
Genomic Map