Assay Details
| Gene Symbol: rfl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066133.7 | NP_498534.2 | 8 - 9 | 1369 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rfl-1| Gene Symbol: rfl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066133.7 | NP_498534.2 | 8 - 9 | 1369 | 99 | |
Set Membership: |
|