Assay Details
| Gene Symbol: dpy-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027560.5 | NP_001022731.1 | 2 - 3 | 272 | 118 | |
| NR_132452.1 | 1 - 2 | 207 | 118 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-31| Gene Symbol: dpy-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027560.5 | NP_001022731.1 | 2 - 3 | 272 | 118 | |
| NR_132452.1 | 1 - 2 | 207 | 118 | |||
Set Membership: |
|