Assay Details
| Gene Symbol: dpy-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027560.5 | NP_001022731.1 | 7 - 8 | 1637 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-31| Gene Symbol: dpy-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027560.5 | NP_001022731.1 | 7 - 8 | 1637 | 89 | |
Set Membership: |
|