Assay Details
| Gene Symbol: dpy-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027560.5 | NP_001022731.1 | 8 - 9 | 1768 | 77 | |
| NR_132452.1 | 7 - 8 | 1596 | 77 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-31| Gene Symbol: dpy-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027560.5 | NP_001022731.1 | 8 - 9 | 1768 | 77 | |
| NR_132452.1 | 7 - 8 | 1596 | 77 | |||
Set Membership: |
|