Assay Details
| Gene Symbol: cwf-19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025954.7 | NP_001021125.1 | 9 - 10 | 1230 | 67 | |
| NM_001025955.5 | NP_001021126.1 | 1 - 2 | 148 | 67 | ||
More Information
Set Membership: |
|

cwf-19L2
Genomic Map