Assay Details
| Gene Symbol: cwf-19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025954.7 | NP_001021125.1 | 8 - 9 | 1059 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cwf-19L2| Gene Symbol: cwf-19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025954.7 | NP_001021125.1 | 8 - 9 | 1059 | 117 | |
Set Membership: |
|