Assay Details
| Gene Symbol: F08F8.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066219.8 | NP_498620.1 | 8 - 9 | 1086 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F08F8.7| Gene Symbol: F08F8.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066219.8 | NP_498620.1 | 8 - 9 | 1086 | 102 | |
Set Membership: |
|