Assay Details
| Gene Symbol: tbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382915.1 | 1 - 2 | 209 | 103 | ||
| GenBank mRNA | AF303249.1 | - | 1 - 2 | 209 | 103 | |
| L07754.1 | - | 1 - 2 | 211 | 103 | ||
More Information
Set Membership: |
|

tbp-1
Genomic Map