Assay Details
| Gene Symbol: alh-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027759.3 | NP_001022930.1 | 1 - 2 | 119 | 125 | |
| NM_001382916.1 | 1 - 2 | 126 | 125 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alh-12| Gene Symbol: alh-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027759.3 | NP_001022930.1 | 1 - 2 | 119 | 125 | |
| NM_001382916.1 | 1 - 2 | 126 | 125 | |||
Set Membership: |
|