Assay Details
| Gene Symbol: mup-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066244.7 | NP_498645.1 | 15 - 16 | 5128 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mup-4| Gene Symbol: mup-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066244.7 | NP_498645.1 | 15 - 16 | 5128 | 82 | |
Set Membership: |
|