Assay Details
| Gene Symbol: mfsd-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066261.5 | NP_498662.1 | 7 - 8 | 1336 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mfsd-6| Gene Symbol: mfsd-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066261.5 | NP_498662.1 | 7 - 8 | 1336 | 96 | |
Set Membership: |
|