Assay Details
| Gene Symbol: cup-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268016.4 | NP_001254945.1 | 3 - 4 | 957 | 115 | |
| GenBank mRNA | AF482952.1 | - | 3 - 4 | 951 | 115 | |
More Information
Set Membership: |
|

cup-5
Genomic Map