Assay Details
| Gene Symbol: cup-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027550.4 | NP_001022721.2 | 5 - 6 | 1288 | 90 | |
| NM_001027551.5 | NP_001022722.2 | 4 - 5 | 1228 | 90 | ||
| NM_001268016.4 | NP_001254945.1 | 4 - 5 | 1240 | 90 | ||
| NM_001382919.1 | 4 - 5 | 1237 | 90 | |||
| GenBank mRNA | AF482952.1 | - | 4 - 5 | 1234 | 90 | |
More Information
Set Membership: |
|

cup-5
Genomic Map