Assay Details
| Gene Symbol: cup-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027550.4 | NP_001022721.2 | 7 - 8 | 1619 | 154 | |
| NM_001027551.5 | NP_001022722.2 | 6 - 7 | 1559 | 154 | ||
| NM_001268016.4 | NP_001254945.1 | 6 - 7 | 1571 | 154 | ||
| NM_001382919.1 | 6 - 7 | 1568 | 154 | |||
| NM_066263.6 | NP_498664.3 | 7 - 8 | 1584 | 154 | ||
| GenBank mRNA | AF482952.1 | - | 6 - 7 | 1565 | 154 | |
More Information
Set Membership: |
|

cup-5
Genomic Map